siRNA Details
| Gene Symbol: Phf21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109690.1 | NP_001103160.1 | 10 | 1213 | ||
| NM_001109691.1 | NP_001103161.1 | 13 | 1369 | |||
| NM_138755.2 | NP_620094.2 | 12 | 1372 | |||
| GenBank mRNA | AB105178.1 | - | 12 | 1278 | ||
| AK037174.1 | - | 13 | 1370 | |||
| AK129424.1 | - | 12 | 2627 | |||
| AK137085.1 | - | 9 | 1180 | |||
| BC019181.1 | - | 12 | 1227 | 5008954 | ||

Phf21a
Genomic Map