siRNA Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147201.2 | NP_671734.1 | 4 | 484 | ||
| GenBank mRNA | AF302138.1 | - | 4 | 483 | ||
| AK046142.1 | - | 4 | 478 | |||
| AK077440.1 | - | 4 | 520 | |||
| AK170501.1 | - | 4 | 500 | |||
| AK178908.1 | - | 4 | 325 | |||
| AK218775.1 | - | 1 | 23 | |||
| BC004756.1 | - | 4 | 454 | 3154089 | ||
| BC018463.1 | - | 4 | 458 | 3598438 | ||

Nrbp1
Genomic Map