siRNA Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147201.2 | NP_671734.1 | 8, 9 | 854 | ||
| GenBank mRNA | AF302138.1 | - | 8, 9 | 853 | ||
| AK046142.1 | - | 9, 10 | 872 | |||
| AK077440.1 | - | 8, 9 | 890 | |||
| AK170501.1 | - | 8, 9 | 870 | |||
| BC004756.1 | - | 8, 9 | 824 | 3154089 | ||
| BC018463.1 | - | 8, 9 | 828 | 3598438 | ||

Nrbp1
Genomic Map