siRNA Details
| Gene Symbol: PCNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001315529.1 | NP_001302458.1 | 31 | 7210 | ||
| NM_006031.5 | NP_006022.3 | 31 | 7048 | |||
| XM_005261124.4 | XP_005261181.1 | 32 | 7100 | |||
| XM_011529594.2 | XP_011527896.1 | 32 | 7148 | |||
| XM_017028362.1 | XP_016883851.1 | 31 | 7067 | |||
| XM_017028363.1 | XP_016883852.1 | 32 | 7269 | |||
| GenBank mRNA | AB007862.3 | - | 31 | 7211 | ||
| AF515282.1 | - | 31 | 6941 | |||
| U52962.1 | - | 32 | 6960 | |||

PCNT
Genomic Map