siRNA Details
| Gene Symbol: Itgb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005608.2 | NP_001005608.2 | 20 | 2725 | ||
| NM_133663.2 | NP_598424.2 | 20 | 2725 | |||
| GenBank mRNA | AK048888.1 | - | 20 | 2609 | ||
| BC059192.1 | - | 16 | 1920 | |||
| L04678.1 | - | 21 | 2569 | |||

Itgb4
Genomic Map