siRNA Details
| Gene Symbol: Lrrc75a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198861.1 | NP_942561.1 | 3 | 567 | ||
| GenBank mRNA | AK146714.1 | - | 3 | 256 | ||
| AK186775.1 | - | 1 | 23 | |||
| BC046404.1 | - | 3 | 307 | |||
| BC053455.1 | - | 3 | 567 | 6513708 | ||

Lrrc75a
Genomic Map