siRNA Details
| Gene Symbol: Lrrc75a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198861.1 | NP_942561.1 | 4 | 822 | ||
| GenBank mRNA | AK146714.1 | - | 4 | 511 | ||
| AK186775.1 | - | 2 | 279 | |||
| BC053455.1 | - | 4 | 822 | 6513708 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc75a| Gene Symbol: Lrrc75a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198861.1 | NP_942561.1 | 4 | 822 | ||
| GenBank mRNA | AK146714.1 | - | 4 | 511 | ||
| AK186775.1 | - | 2 | 279 | |||
| BC053455.1 | - | 4 | 822 | 6513708 | ||