siRNA Details
| Gene Symbol: Trpv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001445.2 | NP_001001445.1 | 8 | 1532 | ||
| GenBank mRNA | AB180097.1 | - | 7 | 1280 | ||
| AJ620495.1 | - | 7 | 1320 | |||
| AK051315.1 | - | 8 | 1390 | |||
| AY445519.1 | - | 7 | 1280 | |||
| AY452083.1 | - | 7 | 1313 | |||
| AY452084.1 | - | 7 | 1283 | |||

Trpv1
Genomic Map