siRNA Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148673.3 | NP_683514.2 | 9 | 980 | ||
| NM_198936.1 | NP_945174.1 | 8 | 901 | |||
| GenBank mRNA | AK029117.1 | - | 8 | 871 | ||
| AK046262.1 | - | 8 | 872 | |||
| AK047597.1 | - | 9 | 968 | |||
| AK049178.1 | - | 8 | 870 | |||
| BC013810.1 | - | 6 | 500 | |||
| BC060954.1 | - | 8 | 869 | |||
| BC082780.1 | - | 9 | 960 | |||
| BC106099.1 | - | 8 | 882 | |||

Slu7
Genomic Map