siRNA Details
| Gene Symbol: BC049762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177567.3 | NP_808235.1 | 2 | 386 | ||
| GenBank mRNA | AK076829.1 | - | 2 | 387 | ||
| AK154967.1 | - | 3 | 570 | |||
| BC049762.1 | - | 2 | 263 | 6774041 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC049762| Gene Symbol: BC049762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177567.3 | NP_808235.1 | 2 | 386 | ||
| GenBank mRNA | AK076829.1 | - | 2 | 387 | ||
| AK154967.1 | - | 3 | 570 | |||
| BC049762.1 | - | 2 | 263 | 6774041 | ||