siRNA Details
| Gene Symbol: Gm13103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177571.3 | NP_808239.2 | 4 | 1442 | ||
| GenBank mRNA | AK054347.1 | - | 4 | 1444 | ||
| BB555205.2 | - | 1 | 241 | |||
| BC096438.1 | - | 4 | 1439 | 30481183 | ||
| Gene Symbol: Gm13088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126325.1 | NP_001119797.1 | 4 | 1434 | ||

Gm13103
Genomic Map