siRNA Details
| Gene Symbol: Gm13023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007077.2 | NP_001007078.2 | 1, 2 | 106 | ||
| GenBank mRNA | AK135972.1 | - | 1, 2 | 89 | ||
| BC084594.1 | - | 1, 2 | 87 | 30478700 | ||
| CA559385.1 | - | 1, 2 | 76 | |||
| CF174133.1 | - | 1, 2 | 114 | |||

Gm13023
Genomic Map