siRNA Details
| Gene Symbol: Rimkla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177572.4 | NP_808240.2 | 2, 3 | 534 | ||
| GenBank mRNA | AK047164.1 | - | 2, 3 | 529 | ||
| AK163360.1 | - | 2, 3 | 525 | |||
| BC057371.1 | - | 2, 3 | 515 | 6835897 | ||
| BU611721.1 | - | 2, 3 | 564 | |||

Rimkla
Genomic Map