siRNA Details
| Gene Symbol: 9930104L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177573.3 | NP_808241.2 | 3, 4 | 748 | ||
| GenBank mRNA | AK158126.1 | - | 3, 4 | 723 | ||
| BC099946.1 | - | 3, 4 | 628 | 30102988 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9930104L06Rik| Gene Symbol: 9930104L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177573.3 | NP_808241.2 | 3, 4 | 748 | ||
| GenBank mRNA | AK158126.1 | - | 3, 4 | 723 | ||
| BC099946.1 | - | 3, 4 | 628 | 30102988 | ||