siRNA Details
| Gene Symbol: Gm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CO797249.1 | - | 1 | 199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5| Gene Symbol: Gm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CO797249.1 | - | 1 | 199 | ||