siRNA Details
| Gene Symbol: Vps37d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199677.1 | NP_001186606.1 | 4 | 1442 | ||
| NM_177574.4 | NP_808242.1 | 2 | 1187 | |||
| GenBank mRNA | AK090181.1 | - | 2 | 1189 | ||
| AK158287.1 | - | 4 | 1371 | |||
| AY081953.2 | - | 4 | 1442 | |||
| AY352519.1 | - | 2 | 1187 | |||
| BC044789.1 | - | 4 | 1345 | 3484221 | ||
| BU704550.1 | - | 1 | 298 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001128193, BC166560.

Vps37d
Genomic Map