siRNA Details
| Gene Symbol: Tet3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183138.2 | NP_898961.2 | 11 | 4034 | ||
| GenBank mRNA | AK084164.1 | - | 6 | 1390 | ||
| BC096437.1 | - | 7 | 1548 | |||
| HQ423151.1 | - | 11 | 4208 | |||
| HQ423152.1 | - | 10 | 4151 | |||
| JX946278.1 | - | 12 | 4740 | |||

Tet3
Genomic Map