siRNA Details
| Gene Symbol: Mical3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153396.3 | NP_700445.2 | 18 | 2785 | ||
| GenBank mRNA | AF536756.1 | - | 18 | 2748 | ||
| AK132669.1 | - | 18 | 2769 | |||
| AK198084.1 | - | 2 | 176 | |||
| BC138257.1 | - | 18 | 2746 | 8861276 | ||
| BC145214.1 | - | 19 | 2832 | |||

Mical3
Genomic Map