siRNA Details
| Gene Symbol: Mical3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270475.1 | NP_001257404.1 | 17 | 2696 | ||
| NM_153396.3 | NP_700445.2 | 17 | 2662 | |||
| GenBank mRNA | AF536756.1 | - | 17 | 2625 | ||
| AK132669.1 | - | 17 | 2646 | |||
| AK198084.1 | - | 1 | 53 | |||
| BC090651.1 | - | 3 | 325 | |||
| BC138257.1 | - | 17 | 2623 | 8861276 | ||
| BC145214.1 | - | 18 | 2709 | |||

Mical3
Genomic Map