siRNA Details
| Gene Symbol: Mical3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270475.1 | NP_001257404.1 | 26 | 4273 | ||
| GenBank mRNA | AK147803.1 | - | 9 | 1537 | ||
| BC043122.1 | - | 7 | 1172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mical3| Gene Symbol: Mical3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270475.1 | NP_001257404.1 | 26 | 4273 | ||
| GenBank mRNA | AK147803.1 | - | 9 | 1537 | ||
| BC043122.1 | - | 7 | 1172 | |||