siRNA Details
| Gene Symbol: Klf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178357.3 | NP_848134.1 | 3 | 925 | ||
| GenBank mRNA | AJ459773.2 | - | 3 | 771 | ||
| AK036618.1 | - | 3 | 911 | |||
| AK038397.1 | - | 3 | 1023 | |||
| AK050156.1 | - | 3 | 927 | |||
| AK054170.1 | - | 3 | 897 | |||
| AK082838.1 | - | 3 | 914 | |||
| AK084913.1 | - | 3 | 898 | |||
| AK085319.1 | - | 3 | 900 | |||
| AK089044.1 | - | 3 | 905 | |||
| AK161233.1 | - | 3 | 917 | |||
| BC060642.1 | - | 3 | 923 | 6850350 | ||

Klf11
Genomic Map