siRNA Details
| Gene Symbol: Tmem199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199199.3 | NP_954669.2 | 2 | 291 | ||
| GenBank mRNA | AK156856.1 | - | 2 | 226 | ||
| AK158039.1 | - | 2 | 255 | |||
| BC050977.1 | - | 2 | 254 | |||
| BC147070.1 | - | 2 | 279 | 9056057 | ||
| BC147071.1 | - | 2 | 279 | 9056065 | ||
| BY255000.1 | - | 2 | 291 | |||

Tmem199
Genomic Map