siRNA Details
| Gene Symbol: Tdpoz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148949.2 | NP_683751.2 | 2 | 520 | ||
| GenBank mRNA | AF290198.1 | - | 2 | 505 | ||
| BC125302.1 | - | 1 | 382 | 40129817 | ||
| BC125304.1 | - | 1 | 382 | 40129819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdpoz1| Gene Symbol: Tdpoz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148949.2 | NP_683751.2 | 2 | 520 | ||
| GenBank mRNA | AF290198.1 | - | 2 | 505 | ||
| BC125302.1 | - | 1 | 382 | 40129817 | ||
| BC125304.1 | - | 1 | 382 | 40129819 | ||