siRNA Details
| Gene Symbol: Hectd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144788.2 | NP_659037.2 | 25 | 4953 | ||
| GenBank mRNA | AK141970.1 | - | 3 | 868 | ||
| AK173106.1 | - | 5 | 1482 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hectd1| Gene Symbol: Hectd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144788.2 | NP_659037.2 | 25 | 4953 | ||
| GenBank mRNA | AK141970.1 | - | 3 | 868 | ||
| AK173106.1 | - | 5 | 1482 | |||