siRNA Details
| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172255.3 | NP_758459.2 | 26, 27 | 3402 | ||
| GenBank mRNA | AK173155.1 | - | 8, 9 | 1047 | ||
| BC037177.1 | - | 26, 27 | 3339 | 4948388 | ||
| BC042568.1 | - | 17, 18 | 3477 | |||

Wdr11
Genomic Map