siRNA Details
| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172255.3 | NP_758459.2 | 24 | 3091 | ||
| GenBank mRNA | AK173155.1 | - | 6 | 736 | ||
| BC037177.1 | - | 24 | 3028 | 4948388 | ||
| BC042568.1 | - | 15 | 3166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr11| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172255.3 | NP_758459.2 | 24 | 3091 | ||
| GenBank mRNA | AK173155.1 | - | 6 | 736 | ||
| BC037177.1 | - | 24 | 3028 | 4948388 | ||
| BC042568.1 | - | 15 | 3166 | |||