siRNA Details
| Gene Symbol: Wdr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172255.3 | NP_758459.2 | 20 | 2716 | ||
| GenBank mRNA | AK043051.1 | - | 18 | 2334 | ||
| AK173155.1 | - | 2 | 361 | |||
| AK188029.1 | - | 2 | 122 | |||
| BC037177.1 | - | 20 | 2653 | 4948388 | ||
| BC042568.1 | - | 13 | 1555 | |||

Wdr11
Genomic Map