siRNA Details
| Gene Symbol: Baiap2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177580.3 | NP_808248.1 | 10 | 1217 | ||
| GenBank mRNA | BC048937.1 | - | 10 | 1155 | 6396012 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baiap2l2| Gene Symbol: Baiap2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177580.3 | NP_808248.1 | 10 | 1217 | ||
| GenBank mRNA | BC048937.1 | - | 10 | 1155 | 6396012 | |