siRNA Details
Gene Symbol: Baiap2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177580.3 | NP_808248.1 | 6 | 559 | ||
GenBank mRNA | AK143783.1 | - | 6 | 544 | ||
BC048937.1 | - | 6 | 497 | 6396012 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Baiap2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177580.3 | NP_808248.1 | 6 | 559 | ||
GenBank mRNA | AK143783.1 | - | 6 | 544 | ||
BC048937.1 | - | 6 | 497 | 6396012 |