siRNA Details
| Gene Symbol: Dtx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172442.3 | NP_766030.3 | 6, 7 | 2146 | ||
| GenBank mRNA | AK086624.1 | - | 5, 6 | 715 | ||
| BC044798.1 | - | 1, 2 | 29 | |||
| BC058647.1 | - | 6, 7 | 1604 | 6405881 | ||
| BC069975.1 | - | 5, 6 | 686 | |||
| CA328050.1 | - | 5, 6 | 583 | |||

Dtx4
Genomic Map