siRNA Details
| Gene Symbol: Camkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199676.1 | NP_001186605.1 | 4, 5 | 784 | ||
| NM_145358.2 | NP_663333.1 | 4, 5 | 784 | |||
| GenBank mRNA | AF453383.1 | - | 3, 4 | 567 | ||
| AK031399.1 | - | 4, 5 | 709 | |||
| AK032070.1 | - | 4, 5 | 757 | |||
| AK044660.1 | - | 4, 5 | 773 | |||
| AK160407.1 | - | 4, 5 | 700 | |||
| BC023103.1 | - | 3, 4 | 617 | 3993474 | ||

Camkk2
Genomic Map