siRNA Details
| Gene Symbol: Cfap69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172447.2 | NP_766035.1 | 22 | 2812 | ||
| GenBank mRNA | AK029553.1 | - | 22 | 2821 | ||
| AK039313.1 | - | 22 | 2813 | |||
| AK076835.1 | - | 22 | 2802 | |||
| AK140211.1 | - | 18 | 2278 | |||
| AK164190.1 | - | 14 | 1757 | |||
| BC050971.1 | - | 12 | 1498 | |||
| BC054079.1 | - | 22 | 2763 | |||
| BC062906.1 | - | 22 | 2777 | 6813710 | ||

Cfap69
Genomic Map