siRNA Details
| Gene Symbol: Gtpbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153116.1 | NP_694756.1 | 8 | 787 | ||
| GenBank mRNA | AK139595.1 | - | 8 | 801 | ||
| AK160252.1 | - | 8 | 826 | |||
| BC034507.1 | - | 8 | 813 | 4222393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtpbp10| Gene Symbol: Gtpbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153116.1 | NP_694756.1 | 8 | 787 | ||
| GenBank mRNA | AK139595.1 | - | 8 | 801 | ||
| AK160252.1 | - | 8 | 826 | |||
| BC034507.1 | - | 8 | 813 | 4222393 | ||