siRNA Details
| Gene Symbol: ADCY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281768.1 | NP_001268697.1 | 7, 8 | 1019 | ||
| NM_021116.2 | NP_066939.1 | 6, 7 | 1333 | |||
| XM_005249584.3 | XP_005249641.1 | 6, 7 | 1331 | |||
| XM_005249585.2 | XP_005249642.1 | 6, 7 | 1331 | |||
| GenBank mRNA | AB209496.1 | - | 6, 7 | 1278 | ||
| L05500.1 | - | 4, 5 | 477 | |||

ADCY1
Genomic Map