siRNA Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172449.2 | NP_766037.2 | 32 | 6529 | ||
| GenBank mRNA | AK051895.1 | - | 10 | 2427 | ||
| AK122329.1 | - | 9 | 1610 | |||
| BC055691.1 | - | 12 | 1280 | |||
| BC066146.1 | - | 10 | 1489 | 30470084 | ||
| BC083181.1 | - | 12 | 1329 | |||

Bzrap1
Genomic Map