siRNA Details
| Gene Symbol: BC034090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078517.1 | - | 2 | 573 | ||
| AK129406.1 | - | 2 | 586 | |||
| BC034090.1 | - | 2 | 415 | |||
| BG086900.2 | - | 3 | 467 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC034090| Gene Symbol: BC034090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078517.1 | - | 2 | 573 | ||
| AK129406.1 | - | 2 | 586 | |||
| BC034090.1 | - | 2 | 415 | |||
| BG086900.2 | - | 3 | 467 | |||