siRNA Details
Gene Symbol: BC034090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078517.1 | - | 2 | 1465 | ||
AK129406.1 | - | 2 | 1475 | |||
BC034090.1 | - | 2 | 1304 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC034090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078517.1 | - | 2 | 1465 | ||
AK129406.1 | - | 2 | 1475 | |||
BC034090.1 | - | 2 | 1304 |