siRNA Details
| Gene Symbol: Fmr1nb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166619.1 | NP_001160091.1 | 1 | 164 | ||
| NM_001166620.1 | NP_001160092.1 | 1 | 164 | |||
| NM_174993.2 | NP_778158.2 | 1 | 164 | |||
| GenBank mRNA | AK076190.1 | - | 1 | 122 | ||
| AY214130.1 | - | 1 | 69 | |||
| BC099411.1 | - | 1 | 164 | 30788714 | ||
| BC138087.1 | - | 1 | 145 | 8861106 | ||
| BC145139.1 | - | 1 | 145 | 9053689 | ||
| BC145140.1 | - | 1 | 145 | 9053690 | ||
| BC171922.1 | - | 1 | 145 | 9053688 | ||
| BC171923.1 | - | 1 | 145 | 9053691 | ||
| BC171947.1 | - | 1 | 145 | 9054233 | ||
| BC171948.1 | - | 1 | 145 | 9054234 | ||

Fmr1nb
Genomic Map