siRNA Details
| Gene Symbol: Esrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290383.1 | NP_001277312.1 | 13 | 2041 | ||
| NM_194055.3 | NP_918944.2 | 13 | 2044 | |||
| GenBank mRNA | AK140848.1 | - | 13 | 2042 | ||
| BC031468.1 | - | 11 | 1335 | 4037840 | ||
| BC059280.1 | - | 11 | 1345 | |||

Esrp1
Genomic Map