siRNA Details
| Gene Symbol: Urb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029497.1 | NP_083773.1 | 26 | 4478 | ||
| GenBank mRNA | AK029516.1 | - | 3 | 307 | ||
| AK220219.1 | - | 5 | 956 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Urb1| Gene Symbol: Urb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029497.1 | NP_083773.1 | 26 | 4478 | ||
| GenBank mRNA | AK029516.1 | - | 3 | 307 | ||
| AK220219.1 | - | 5 | 956 | |||