siRNA Details
Gene Symbol: Alg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243161.1 | NP_001230090.1 | 3 | 975 | ||
NM_183142.4 | NP_898965.1 | 3 | 849 | |||
GenBank mRNA | AB288237.1 | - | 3 | 969 | ||
AB288238.1 | - | 3 | 973 | |||
AK046475.1 | - | 3 | 851 | |||
AK134312.1 | - | 3 | 950 | |||
AK157746.1 | - | 3 | 975 | |||
BC061469.1 | - | 3 | 958 | 6821552 |
