siRNA Details
| Gene Symbol: Vcpkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033236.2 | NP_001028408.2 | 5 | 646 | ||
| GenBank mRNA | AK083148.1 | - | 1 | 3046 | ||
| AK144832.1 | - | 5 | 609 | |||
| BC138942.1 | - | 5 | 728 | 8861964 | ||
| BC138943.1 | - | 5 | 728 | 8861965 | ||
| BC145471.1 | - | 5 | 717 | |||

Vcpkmt
Genomic Map