siRNA Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029838.2 | NP_001025009.1 | 8 | 976 | ||
| NM_148950.3 | NP_683752.2 | 7 | 871 | |||
| GenBank mRNA | AB086817.1 | - | 5 | 697 | ||
| AF487460.1 | - | 6 | 777 | |||
| AK082952.1 | - | 8 | 921 | |||
| BC050865.1 | - | 7 | 805 | 6332968 | ||

Pknox2
Genomic Map