siRNA Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085498.2 | NP_001078967.1 | 6 | 549 | ||
| GenBank mRNA | AK162346.1 | - | 2 | 126 | ||
| AK172123.1 | - | 5 | 698 | |||
| AK184598.1 | - | 1 | 49 | |||
| BC147754.1 | - | 6 | 544 | 9056703 | ||
| BC147755.1 | - | 6 | 544 | 9056704 | ||
| BG078687.2 | - | 6 | 464 | |||

Chmp6
Genomic Map