siRNA Details
| Gene Symbol: Aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177583.4 | NP_808251.1 | 2 | 249 | ||
| GenBank mRNA | AK028521.1 | - | 2 | 263 | ||
| AK049828.1 | - | 2 | 250 | |||
| AK154847.1 | - | 2 | 317 | |||
| AK162585.1 | - | 2 | 231 | |||
| BC079659.1 | - | 2 | 281 | 6853991 | ||

Aph1b
Genomic Map