siRNA Details
| Gene Symbol: Aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177583.4 | NP_808251.1 | 6 | 1552 | ||
| GenBank mRNA | AK028521.1 | - | 7 | 1811 | ||
| AK049828.1 | - | 6 | 1553 | |||
| AK154847.1 | - | 6 | 1620 | |||
| AK162585.1 | - | 7 | 1787 | |||
| BC079659.1 | - | 6 | 1584 | 6853991 | ||

Aph1b
Genomic Map