siRNA Details
| Gene Symbol: Tmem132b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190352.1 | NP_001177281.1 | 2 | 558 | ||
| GenBank mRNA | AK220418.1 | - | 1 | 258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem132b| Gene Symbol: Tmem132b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190352.1 | NP_001177281.1 | 2 | 558 | ||
| GenBank mRNA | AK220418.1 | - | 1 | 258 | ||