siRNA Details
| Gene Symbol: Slc9c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198106.4 | NP_932774.3 | 18 | 2499 | ||
| GenBank mRNA | AK131702.1 | - | 1 | 40 | ||
| AY368685.1 | - | 18 | 2459 | |||
| BC145031.1 | - | 2106 | ||||
| BC145032.1 | - | 2321 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9c1| Gene Symbol: Slc9c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198106.4 | NP_932774.3 | 18 | 2499 | ||
| GenBank mRNA | AK131702.1 | - | 1 | 40 | ||
| AY368685.1 | - | 18 | 2459 | |||
| BC145031.1 | - | 2106 | ||||
| BC145032.1 | - | 2321 | ||||