siRNA Details
| Gene Symbol: Slc9c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198106.4 | NP_932774.3 | 20, 21 | 2791 | ||
| GenBank mRNA | AK131702.1 | - | 3, 4 | 332 | ||
| AY368685.1 | - | 20, 21 | 2751 | |||
| BC145031.1 | - | 2398 | ||||
| BC145032.1 | - | 2613 | ||||

Slc9c1
Genomic Map