siRNA Details
| Gene Symbol: Phldb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252442.1 | NP_001239371.1 | 2 | 1324 | ||
| NM_153412.4 | NP_700461.2 | 2 | 1324 | |||
| GenBank mRNA | AF506821.1 | - | 2 | 1325 | ||
| AK049595.1 | - | 2 | 1178 | |||
| AK050493.1 | - | 3 | 1491 | |||
| AK165251.1 | - | 2 | 1286 | |||
| BC050915.1 | - | 2 | 1180 | 6405379 | ||
| BC060683.1 | - | 2 | 1180 | 6405379 | ||

Phldb2
Genomic Map